Assay Details
Target Gene Details
Entrez Gene ID: | 5521 |
Gene Name: | protein phosphatase 2 regulatory subunit Bbeta |
Gene Aliases: |
B55BETA, PP2AB55BETA, PP2ABBETA, PP2APR55B, PP2APR55BETA, PR2AB55BETA, PR2ABBETA, PR2APR55BETA, PR52B, PR55-BETA, PR55BETA, SCA12 |
Location: |
Chr.5:146589504-147081520 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP2R2B | NM_001271899.1 | NP_001258828.1 | ||
| NM_001271900.1 | NP_001258829.1 | |||
| NM_181674.2 | NP_858060.2 | |||
| NM_181676.2 | NP_858062.1 | |||
| NM_181677.2 | NP_858063.1 | |||
| NM_181678.2 | NP_858064.1 | |||
| AK294659.1 | ||||
| AK295347.1 | ||||
| AV726282.1 | ||||
| BC031790.1 | AAH31790.1 | |||
| BI669304.1 | ||||
| DA155307.1 | ||||
| DA365141.1 | ||||
| DA522334.1 |
Target Gene Details
Entrez Gene ID: | 100874361 |
Gene Name: | PPP2R2B intronic transcript 1 |
Gene Aliases: |
- |
Location: |
Chr.5:146914207-146919506 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP2R2B-IT1 | NR_047115.1 | |||
| BX113093.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv599938 | Chr.5:146789948 - 147426802 on Build GRCh38 | Gain |
|
| nsv519847 | Chr.5:146786815 - 147433084 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map