Assay Details
Target Gene Details
Entrez Gene ID: | 29093 |
Gene Name: | mitochondrial ribosomal protein L22 |
Gene Aliases: |
HSPC158, L22mt, MRP-L22, MRP-L25, RPML25 |
Location: |
Chr.5:154941073-154969411 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL22 | NM_001014990.2 | NP_001014990.1 | ||
| NM_014180.3 | NP_054899.2 | |||
| AF161507.1 | AAF29122.1 | |||
| AK000601.1 | BAA91282.1 | |||
| AK302784.1 | ||||
| BC012565.1 | AAH12565.1 | |||
| BM662504.1 | ||||
| BP372420.1 | ||||
| BU943770.1 | ||||
| CR933679.1 | CAI45973.1 |
More Information
Set Membership: |
|

Genomic Map