Assay Details
Target Gene Details
Entrez Gene ID: | 2668 |
Gene Name: | glial cell derived neurotrophic factor |
Gene Aliases: |
ATF1, ATF2, HFB1-GDNF, HSCR3 |
Location: |
Chr.5:37812677-37840044 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GDNF | NM_000514.3 | NP_000505.1 | ||
| XM_011514030.2 | XP_011512332.1 | |||
| XM_017009337.1 | XP_016864826.1 | |||
| AF053748.1 | AAD43139.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3604748 | Chr.5:37720912 - 37848930 on Build GRCh38 | Gain |
|
| dgv1593e212 | Chr.5:37832461 - 37848988 on Build GRCh38 | Loss |
|
| esv3604751 | Chr.5:37835063 - 37838062 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map