Assay Details
Target Gene Details
Entrez Gene ID: | 64979 |
Gene Name: | mitochondrial ribosomal protein L36 |
Gene Aliases: |
BRIP1, L36mt, MRP-L36, PRPL36, RPMJ |
Location: |
Chr.5:1798385-1801432 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL36 | XM_011514080.2 | XP_011512382.1 | ||
| XM_017009751.1 | XP_016865240.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3603817 | Chr.5:1638442 - 1833414 on Build GRCh38 | Gain |
|
| nsv1033147 | Chr.5:1717188 - 1815707 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map