Assay Details
Target Gene Details
Entrez Gene ID: | 115123 |
Gene Name: | membrane associated ring-CH-type finger 3 |
Gene Aliases: |
MARCH-III, RNF173 |
Location: |
Chr.5:126867714-127032402 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MARCH3 | NM_178450.4 | NP_848545.1 | ||
| XM_011543128.2 | XP_011541430.1 | |||
| XM_011543129.1 | XP_011541431.1 | |||
| XM_011543130.1 | XP_011541432.1 | |||
| XM_011543131.2 | XP_011541433.1 | |||
| XM_017009014.1 | XP_016864503.1 | |||
| AF055007.1 | ||||
| AK290129.1 | ||||
| BC047569.1 | AAH47569.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1146007 | Chr.5:126890607 - 126896708 on Build GRCh38 | Duplication |
|
| esv3570411 | Chr.5:126890063 - 126903760 on Build GRCh38 | Loss |
|
| esv3606695 | Chr.5:126894710 - 126905929 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map