Assay Details
Target Gene Details
Entrez Gene ID: | 11044 |
Gene Name: | PAP associated domain containing 7 |
Gene Aliases: |
LAK-1, LAK1, POLK, POLS, TRF4, TRF4-1, TRF41, TUTASE5 |
Location: |
Chr.5:6713952-6757048 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAPD7 | NM_001171805.1 | NP_001165276.1 | ||
| NM_006999.4 | NP_008930.1 | |||
| XM_005248234.3 | XP_005248291.1 | |||
| XM_017008989.1 | XP_016864478.1 | |||
| AF089896.1 | AAD45198.1 | |||
| AI871781.1 | ||||
| BC084567.1 | ||||
| KC424495.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv461922 | Chr.5:6680002 - 6744685 on Build GRCh38 | Loss |
|
| nsv822995 | Chr.5:6708036 - 6717215 on Build GRCh38 | Gain |
|
| nsv596958 | Chr.5:6712921 - 6715972 on Build GRCh38 | Gain+Loss |
|
| nsv822994 | Chr.5:6701714 - 6766012 on Build GRCh38 | Gain |
|
| nsv596949 | Chr.5:6705744 - 6777656 on Build GRCh38 | Loss |
|
| nsv596965 | Chr.5:6713146 - 6719051 on Build GRCh38 | Loss |
|
| nsv822993 | Chr.5:6689024 - 6753962 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map