Assay Details
Target Gene Details
Entrez Gene ID: | 9315 |
Gene Name: | neuronal regeneration related protein |
Gene Aliases: |
C5orf13, D4S114, P311, PRO1873, PTZ17, SEZ17 |
Location: |
Chr.5:111729303-111976931 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NREP | NM_001142474.1 | NP_001135946.1 | ||
| NM_001142475.1 | NP_001135947.1 | |||
| AK298779.1 | ||||
| DA027062.1 | ||||
| DA694580.1 | ||||
| DC365764.1 |
Target Gene Details
Entrez Gene ID: | 100873948 |
Gene Name: | NREP antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.5:111912508-112017306 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NREP-AS1 | NR_046678.1 | |||
| BX114072.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv599367 | Chr.5:111588583 - 112267016 on Build GRCh38 | Gain |
|
| esv3606281 | Chr.5:111929634 - 111999912 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map