Assay Details
Target Gene Details
Entrez Gene ID: | 105379194 |
Gene Name: | uncharacterized LOC105379194 |
Gene Aliases: |
- |
Location: |
Chr.5:138744430-138753924 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105379194 | NR_134244.1 | |||
| NR_134245.1 | ||||
| AI792600.1 | ||||
| HY038745.1 | ||||
| HY224594.1 |

Genomic Map