Assay Details
Target Gene Details
Entrez Gene ID: | 2890 |
Gene Name: | glutamate ionotropic receptor AMPA type subunit 1 |
Gene Aliases: |
GLUH1, GLUR1, GLURA, GluA1, HBGR1 |
Location: |
Chr.5:153489615-153813873 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRIA1 | XM_011537635.2 | XP_011535937.1 | ||
| AK310189.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1019051 | Chr.5:153171303 - 153678336 on Build GRCh38 | Gain |
|
| esv2657105 | Chr.5:153385677 - 153727589 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map