Assay Details
Target Gene Details
Entrez Gene ID: | 2890 |
Gene Name: | glutamate ionotropic receptor AMPA type subunit 1 |
Gene Aliases: |
GLUH1, GLUR1, GLURA, GluA1, HBGR1 |
Location: |
Chr.5:153489615-153813873 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRIA1 | NM_000827.3 | NP_000818.2 | ||
| NM_001114183.1 | NP_001107655.1 | |||
| NM_001258019.1 | NP_001244948.1 | |||
| NM_001258020.1 | NP_001244949.1 | |||
| NM_001258021.1 | NP_001244950.1 | |||
| NM_001258022.1 | NP_001244951.1 | |||
| NM_001258023.1 | NP_001244952.1 | |||
| NR_047578.1 | ||||
| XM_011537635.2 | XP_011535937.1 | |||
| XM_017009392.1 | XP_016864881.1 | |||
| XM_017009393.1 | XP_016864882.1 | |||
| AB209094.1 | BAD92331.1 | |||
| AK289565.1 | ||||
| AK294011.1 | ||||
| AK295039.1 | ||||
| AK295184.1 | ||||
| AK295827.1 | ||||
| AK310189.1 | ||||
| AK315934.1 | ||||
| BC111734.1 | AAI11735.1 | |||
| M64752.1 | AAA58613.1 | |||
| M81886.1 | AAA58395.1 | |||
| X58633.1 | CAA41491.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1019051 | Chr.5:153171303 - 153678336 on Build GRCh38 | Gain |
|
| esv2657105 | Chr.5:153385677 - 153727589 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map