Assay Details
Target Gene Details
Entrez Gene ID: | 133686 |
Gene Name: | NAD kinase 2, mitochondrial |
Gene Aliases: |
C5orf33, DECRD, MNADK, NADKD1 |
Location: |
Chr.5:36192589-36242279 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NADK2 | NM_001085411.2 | NP_001078880.1 | ||
| NM_001287340.1 | NP_001274269.1 | |||
| NM_001287341.1 | NP_001274270.1 | |||
| NM_153013.4 | NP_694558.1 | |||
| XM_005248241.4 | XP_005248298.1 | |||
| AK055158.1 | BAB70864.1 | |||
| AK057950.1 | ||||
| AK304009.1 | ||||
| AY360463.1 | AAQ62967.1 | |||
| BC032912.1 | AAH32912.1 | |||
| BC062567.1 | AAH62567.1 | |||
| BX648388.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3604707 | Chr.5:35513484 - 36501574 on Build GRCh38 | Gain |
|
| nsv830262 | Chr.5:36074209 - 36293770 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map