Assay Details
Target Gene Details
Entrez Gene ID: | 501 |
Gene Name: | aldehyde dehydrogenase 7 family member A1 |
Gene Aliases: |
ATQ1, EPD, PDE |
Location: |
Chr.5:126541841-126595390 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALDH7A1 | NM_001182.4 | 18 | 3900 | NP_001173.2 |
| NM_001201377.1 | 18 | 3900 | NP_001188306.1 | |
| NM_001202404.1 | 16 | 3708 | NP_001189333.2 | |
| AK123774.1 | 1 | 258 | ||
| BG680326.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3606679 | Chr.5:126424914 - 126763280 on Build GRCh38 | Gain |
|
| esv3606678 | Chr.5:126424315 - 126543577 on Build GRCh38 | Gain |
|
| esv3606680 | Chr.5:126475605 - 126628219 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map