Assay Details
Target Gene Details
Entrez Gene ID: | 54622 |
Gene Name: | ADP ribosylation factor like GTPase 15 |
Gene Aliases: |
ARFRP2 |
Location: |
Chr.5:53883942-54310590 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARL15 | NM_019087.2 | NP_061960.1 | ||
| XM_011543498.2 | XP_011541800.1 | |||
| XM_011543499.2 | XP_011541801.1 | |||
| XM_011543500.2 | XP_011541802.1 | |||
| XM_017009598.1 | XP_016865087.1 | |||
| AK000058.1 | BAA90915.1 | |||
| AK290886.1 | ||||
| AK311482.1 | ||||
| BC026093.1 | AAH26093.1 | |||
| CR457225.1 | CAG33506.1 |
Target Gene Details
Entrez Gene ID: | 693166 |
Gene Name: | microRNA 581 |
Gene Aliases: |
MIRN581, hsa-mir-581 |
Location: |
Chr.5:53951504-53951599 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR581 | NR_030307.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5666n100 | Chr.5:53231175 - 54453241 on Build GRCh38 | Gain |
|
| nsv830299 | Chr.5:53911190 - 54090770 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map