Assay Details
Target Gene Details
Entrez Gene ID: | 80006 |
Gene Name: | trafficking protein particle complex 13 |
Gene Aliases: |
C5orf44 |
Location: |
Chr.5:65624731-65666136 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRAPPC13 | NM_001093755.1 | 1 | 92 | NP_001087224.1 |
| NM_001093756.1 | 1 | 92 | NP_001087225.1 | |
| NM_001243737.1 | 1 | 92 | NP_001230666.1 | |
| NM_024941.3 | 1 | 92 | NP_079217.2 | |
| NR_003545.1 | 1 | 92 | ||
| XM_017009890.1 | 1 | 46 | XP_016865379.1 | |
| CD105043.1 | 1 | 61 | ||
| DA903408.1 | 1 | 92 | ||
| DB145758.1 | 1 | 90 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3894204 | Chr.5:65586109 - 65714937 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map