Assay Details
Target Gene Details
Entrez Gene ID: | 51780 |
Gene Name: | lysine demethylase 3B |
Gene Aliases: |
5qNCA, C5orf7, JMJD1B, NET22 |
Location: |
Chr.5:138352596-138437027 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KDM3B | NM_016604.3 | NP_057688.2 | ||
| XM_005272018.4 | XP_005272075.1 | |||
| XM_011543489.2 | XP_011541791.1 | |||
| AB029005.2 | ||||
| AF338242.1 | AAK13499.1 | |||
| BC146788.1 | ||||
| BU506660.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1033604 | Chr.5:138291351 - 138404936 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72803845] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map