Assay Details
Target Gene Details
Entrez Gene ID: | 105379194 |
Gene Name: | uncharacterized LOC105379194 |
Gene Aliases: |
- |
Location: |
Chr.5:138744430-138753924 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105379194 | NR_134244.1 | |||
| NR_134245.1 | ||||
| AI792600.1 | ||||
| HY038745.1 | ||||
| HY224594.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs76089690] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map