Assay Details
Target Gene Details
Entrez Gene ID: | 408187 |
Gene Name: | serine peptidase inhibitor, Kazal type 14 (putative) |
Gene Aliases: |
SPINK5L2 |
Location: |
Chr.5:148146372-148176076 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPINK14 | XM_017009467.1 | XP_016864956.1 | ||
| XM_017009468.1 | XP_016864957.1 | |||
| JF509944.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3607101 | Chr.5:148020480 - 148213612 on Build GRCh38 | Loss |
|
| esv3607102 | Chr.5:148026584 - 148359140 on Build GRCh38 | Loss |
|
| nsv830515 | Chr.5:148081289 - 148222684 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map