Assay Details
Target Gene Details
Entrez Gene ID: | 324 |
Gene Name: | APC, WNT signaling pathway regulator |
Gene Aliases: |
BTPS2, DP2, DP2.5, DP3, GS, PPP1R46 |
Location: |
Chr.5:112707505-112846239 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APC | NM_001127511.2 | NP_001120983.2 | ||
| AK294544.1 | ||||
| BP230350.1 | ||||
| DA777787.1 |
More Information
Set Membership: |
|

Genomic Map