Assay Details
Target Gene Details
Entrez Gene ID: | 91937 |
Gene Name: | T-cell immunoglobulin and mucin domain containing 4 |
Gene Aliases: |
SMUCKLER, TIM4 |
Location: |
Chr.5:156915750-156963255 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TIMD4 | XM_017010021.1 | 7 | 3527 | XP_016865510.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3607302 | Chr.5:156839545 - 157055054 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map