Assay Details
Target Gene Details
Entrez Gene ID: | 6715 |
Gene Name: | steroid 5 alpha-reductase 1 |
Gene Aliases: |
S5AR 1 |
Location: |
Chr.5:6633322-6669562 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRD5A1 | NM_001047.3 | NP_001038.1 | ||
| NM_001324322.1 | NP_001311251.1 | |||
| NM_001324323.1 | NP_001311252.1 | |||
| NR_136739.1 | ||||
| AF052126.1 | AAC28620.1 | |||
| AF113128.1 | AAF14869.1 | |||
| AK297208.1 | ||||
| AK297511.1 | ||||
| AK313070.1 | ||||
| AK315996.1 | ||||
| BC007033.1 | AAH07033.1 | |||
| BC008673.2 | AAH08673.1 | |||
| BT006834.1 | AAP35480.1 | |||
| M32313.1 | AAA35490.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4696 | Chr.5:6639024 - 6673657 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map