Assay Details
Target Gene Details
Entrez Gene ID: | 29958 |
Gene Name: | dimethylglycine dehydrogenase |
Gene Aliases: |
DMGDHD, ME2GLYDH |
Location: |
Chr.5:78997564-79069674 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DMGDH | NM_013391.3 | NP_037523.2 | ||
| AF111858.1 | AAF21941.1 | |||
| AK057641.1 | ||||
| AK314736.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762527 | Chr.5:78625376 - 79252373 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map