Assay Details
Target Gene Details
Entrez Gene ID: | 23635 |
Gene Name: | single stranded DNA binding protein 2 |
Gene Aliases: |
HSPC116, SOSS-B2 |
Location: |
Chr.5:81417360-81751253 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SSBP2 | NM_001256732.1 | 17 | 3592 | NP_001243661.1 |
| NM_001256733.1 | 16 | 3508 | NP_001243662.1 | |
| NM_001256734.1 | 16 | 3502 | NP_001243663.1 | |
| NM_001256735.1 | 16 | 3478 | NP_001243664.1 | |
| NM_001256736.1 | 15 | 3379 | NP_001243665.1 | |
| NM_012446.3 | 17 | 3568 | NP_036578.2 | |
| XM_011543297.2 | 16 | 3408 | XP_011541599.1 | |
| XM_017009304.1 | 17 | 3410 | XP_016864793.1 | |
| XM_017009305.1 | 18 | 3499 | XP_016864794.1 | |
| XM_017009306.1 | 16 | 3384 | XP_016864795.1 | |
| XM_017009308.1 | 18 | 3671 | XP_016864797.1 | |
| XM_017009309.1 | 17 | 3735 | XP_016864798.1 | |
| XM_017009310.1 | 17 | 3883 | XP_016864799.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3605594 | Chr.5:81406976 - 81473957 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map