Assay Details
Target Gene Details
Entrez Gene ID: | 103689846 |
Gene Name: | ADAMTS19 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.5:129459559-129460689 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAMTS19-AS1 | NR_125748.1 | 2 | 668 | |
| DA728271.1 | 2 | 668 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830486 | Chr.5:129399335 - 129592170 on Build GRCh38 | Gain |
|
| esv2763498 | Chr.5:129423618 - 130021576 on Build GRCh38 | Gain |
|
| nsv599704 | Chr.5:129170896 - 130297608 on Build GRCh38 | Gain |
|
| esv2752060 | Chr.5:129382408 - 129504408 on Build GRCh38 | Gain |
|
| esv2759377 | Chr.5:129260045 - 129499159 on Build GRCh38 | Loss |
|

Genomic Map