Assay Details
Target Gene Details
Entrez Gene ID: | 115123 |
Gene Name: | membrane associated ring-CH-type finger 3 |
Gene Aliases: |
MARCH-III, RNF173 |
Location: |
Chr.5:126867714-127032402 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MARCH3 | NM_178450.4 | NP_848545.1 | ||
| XM_011543128.2 | XP_011541430.1 | |||
| XM_011543129.1 | XP_011541431.1 | |||
| XM_011543130.1 | XP_011541432.1 | |||
| XM_011543131.2 | XP_011541433.1 | |||
| XM_017009014.1 | XP_016864503.1 | |||
| AF055007.1 | ||||
| AK290129.1 | ||||
| BC047569.1 | AAH47569.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs74922997] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map