Assay Details
Target Gene Details
Entrez Gene ID: | 4208 |
Gene Name: | myocyte enhancer factor 2C |
Gene Aliases: |
C5DELq14.3, DEL5q14.3 |
Location: |
Chr.5:88718241-88904105 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MEF2C | NM_001131005.2 | NP_001124477.1 | ||
| NM_001193347.1 | NP_001180276.1 | |||
| XM_005248511.2 | XP_005248568.1 | |||
| XM_006714619.2 | XP_006714682.1 | |||
| XM_011543396.2 | XP_011541698.1 | |||
| XM_017009480.1 | XP_016864969.1 | |||
| XM_017009481.1 | XP_016864970.1 | |||
| AL833268.1 | ||||
| AL833274.1 | ||||
| BP231922.1 |
Target Gene Details
Entrez Gene ID: | 101929423 |
Gene Name: | MEF2C antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.5:88883330-89467581 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MEF2C-AS1 | NR_109941.1 | |||
| NR_136218.1 | ||||
| NR_136219.1 | ||||
| BG503445.1 | ||||
| BU599131.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv471022 | Chr.5:88769313 - 88955319 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map