Assay Details
Target Gene Details
Entrez Gene ID: | 134526 |
Gene Name: | acyl-CoA thioesterase 12 |
Gene Aliases: |
CACH-1, Cach, STARD15, THEAL |
Location: |
Chr.5:81309045-81394174 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACOT12 | XM_017009045.1 | XP_016864534.1 | ||
| XM_017009046.1 | XP_016864535.1 | |||
| XM_017009047.1 | XP_016864536.1 | |||
| XM_017009048.1 | XP_016864537.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3890664 | Chr.5:81194463 - 81323227 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79781012] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map