Assay Details
Target Gene Details
Entrez Gene ID: | 401190 |
Gene Name: | regulator of G-protein signaling 7 binding protein |
Gene Aliases: |
R7BP |
Location: |
Chr.5:64504565-64614661 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RGS7BP | NM_001029875.2 | NP_001025046.1 | ||
| XM_005248502.3 | XP_005248559.1 | |||
| AK294808.1 | ||||
| AK296443.1 | ||||
| BC140707.1 | ||||
| BX640900.1 | CAE45947.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4861 | Chr.5:64484259 - 64529368 on Build GRCh38 | Deletion |
|
| nsv830324 | Chr.5:64457623 - 64686227 on Build GRCh38 | Loss |
|
| nsv830323 | Chr.5:64375894 - 64568227 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78999271] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map