Assay Details
Target Gene Details
Entrez Gene ID: | 1432 |
Gene Name: | mitogen-activated protein kinase 14 |
Gene Aliases: |
CSBP, CSBP1, CSBP2, CSPB1, EXIP, Mxi2, PRKM14, PRKM15, RK, SAPK2A, p38, p38ALPHA |
Location: |
Chr.6:36027635-36122964 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAPK14 | NM_001315.2 | NP_001306.1 | ||
| NM_139012.2 | NP_620581.1 | |||
| NM_139013.2 | NP_620582.1 | |||
| NM_139014.2 | NP_620583.1 | |||
| XM_006714998.3 | XP_006715061.1 | |||
| XM_011514310.2 | XP_011512612.2 | |||
| XM_017010299.1 | XP_016865788.1 | |||
| XM_017010300.1 | XP_016865789.1 | |||
| XM_017010301.1 | XP_016865790.1 | |||
| XM_017010302.1 | XP_016865791.1 | |||
| XM_017010303.1 | XP_016865792.1 | |||
| XM_017010304.1 | XP_016865793.1 | |||
| AB074150.1 | BAB85654.1 | |||
| AF100544.1 | AAF36770.1 | |||
| AK291709.1 | ||||
| AK303414.1 | ||||
| BC000092.1 | AAH00092.1 | |||
| BC031574.1 | AAH31574.1 | |||
| BT006933.1 | AAP35579.1 | |||
| CR536505.1 | CAG38743.1 | |||
| FJ032367.1 | ||||
| FJ032368.1 | ||||
| L35263.1 | AAA57455.1 | |||
| L35264.1 | AAA57456.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528432 | Chr.6:35991094 - 36067577 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72657684] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map