Assay Details
Target Gene Details
Entrez Gene ID: | 5269 |
Gene Name: | serpin family B member 6 |
Gene Aliases: |
CAP, DFNB91, MSTP057, PI-6, PI6, PTI, SPI3 |
Location: |
Chr.6:2948159-2972165 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950760 | Chr.6:2946367 - 2957166 on Build GRCh38 | Deletion |
|
| nsv600832 | Chr.6:2945097 - 2970334 on Build GRCh38 | Loss |
|
| nsv830575 | Chr.6:2847376 - 3035682 on Build GRCh38 | Loss |
|
| nsv1018369 | Chr.6:2944928 - 2961123 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map