Assay Details
Target Gene Details
Entrez Gene ID: | 29078 |
Gene Name: | NADH:ubiquinone oxidoreductase complex assembly factor 4 |
Gene Aliases: |
C6orf66, HRPAP20, HSPC125, My013, bA22L21.1 |
Location: |
Chr.6:96889311-96897891 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFAF4 | NM_014165.3 | NP_054884.1 | ||
| AF060508.1 | AAG43126.1 | |||
| AF161474.1 | AAF29089.1 | |||
| AK026245.1 | ||||
| AK311850.1 | ||||
| BC039464.1 | AAH39464.1 | |||
| DB253189.1 |
More Information
Set Membership: |
|

Genomic Map