Assay Details
Target Gene Details
Entrez Gene ID: | 100874531 |
Gene Name: | long intergenic non-protein coding RNA 581 |
Gene Aliases: |
linc-MBOAT1-4 |
Location: |
Chr.6:21486061-21511892 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC00581 | NR_103790.1 | |||
| BX118339.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs73737531] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map