Assay Details
Target Gene Details
Entrez Gene ID: | 285755 |
Gene Name: | peptidylprolyl isomerase like 6 |
Gene Aliases: |
PPIase, RSPH12, bA425D10.6, dJ919F19.1 |
Location: |
Chr.6:109390215-109441171 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPIL6 | NM_001111298.2 | 9 | 2959 | NP_001104768.2 |
| NM_001286360.1 | 7 | 2785 | NP_001273289.1 | |
| NM_001286361.1 | 6 | 2649 | NP_001273290.1 | |
| NM_173672.4 | 8 | 2881 | NP_775943.1 | |
| NR_104429.1 | 9 | 3001 |
More Information
Set Membership: |
|

Genomic Map