Assay Details
Target Gene Details
Entrez Gene ID: | 9457 |
Gene Name: | four and a half LIM domains 5 |
Gene Aliases: |
1700027G07Rik, ACT, FHL-5, dJ393D12.2 |
Location: |
Chr.6:96562525-96618626 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FHL5 | NM_001170807.2 | NP_001164278.1 | ||
| NM_001322466.1 | NP_001309395.1 | |||
| NM_001322467.1 | NP_001309396.1 | |||
| NM_020482.5 | NP_065228.4 | |||
| AF278541.1 | AAF85978.1 | |||
| AK057611.1 | ||||
| AK058175.1 | ||||
| AK223539.1 | BAD97259.1 | |||
| AK314608.1 | ||||
| BC021723.2 | AAH21723.1 | |||
| BC029490.1 | AAH29490.1 | |||
| DA099831.1 | ||||
| DA234984.1 | ||||
| HY020942.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs113292035] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map