Assay Details
Target Gene Details
Entrez Gene ID: | 9532 |
Gene Name: | BCL2 associated athanogene 2 |
Gene Aliases: |
BAG-2, dJ417I1.2 |
Location: |
Chr.6:57172306-57185215 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BAG2 | NM_004282.3 | NP_004273.1 | ||
| XM_005249490.3 | XP_005249547.1 | |||
| XM_011514998.2 | XP_011513300.1 | |||
| XM_011514999.2 | XP_011513301.1 | |||
| AF095192.1 | AAD16121.1 | |||
| AK001015.1 | ||||
| AK023735.1 | ||||
| AK301934.1 | ||||
| AL050287.1 | CAB43388.1 | |||
| BC125039.1 | ||||
| CR533496.1 | CAG38527.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv603190 | Chr.6:57109104 - 57234175 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map