Assay Details
Target Gene Details
Entrez Gene ID: | 51660 |
Gene Name: | mitochondrial pyruvate carrier 1 |
Gene Aliases: |
BRP44L, CGI-129, MPYCD, dJ68L15.3 |
Location: |
Chr.6:166364919-166383013 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MPC1 | NM_001270879.1 | NP_001257808.1 | ||
| NM_016098.3 | NP_057182.1 | |||
| NR_073093.1 | ||||
| AF125101.1 | AAD39918.1 | |||
| AF151887.1 | AAD34124.1 | |||
| AK310934.1 | ||||
| AK312201.1 | ||||
| AL556427.3 | ||||
| AL561589.3 | ||||
| BC000810.1 | AAH00810.1 | |||
| BF665465.1 | ||||
| BG828174.1 | ||||
| DC402302.1 |
More Information
Set Membership: |
|

Genomic Map