Assay Details
Target Gene Details
Entrez Gene ID: | 10591 |
Gene Name: | 2'-deoxynucleoside 5'-phosphate N-hydrolase 1 |
Gene Aliases: |
C6orf108, RCL, dJ330M21.3 |
Location: |
Chr.6:43225629-43229473 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DNPH1 | NM_006443.2 | NP_006434.1 | ||
NM_199184.1 | NP_954653.1 | |||
AF040105.1 | AAB96766.1 | |||
BC011683.2 | AAH11683.1 | |||
BM747628.1 | ||||
EU585603.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv602997 | Chr.6:43221202 - 43291349 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv1017398 | Chr.6:42659705 - 43400998 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
