Assay Details
Target Gene Details
Entrez Gene ID: | 5108 |
Gene Name: | pericentriolar material 1 |
Gene Aliases: |
PTC4, RET/PCM-1 |
Location: |
Chr.8:17922763-18029948 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv7121n100 | Chr.8:17766079 - 17957752 on Build GRCh38 | Gain |
|
| nsv610701 | Chr.8:17787519 - 17956578 on Build GRCh38 | Gain |
|
| esv3616500 | Chr.8:17771011 - 17959017 on Build GRCh38 | Gain |
|
| esv2762735 | Chr.8:17723646 - 17957764 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs80062473] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map