Assay Details
Target Gene Details
Entrez Gene ID: | 7994 |
Gene Name: | lysine acetyltransferase 6A |
Gene Aliases: |
MOZ, MRD32, MYST-3, MYST3, RUNXBP2, ZC2HC6A, ZNF220 |
Location: |
Chr.8:41929479-42052026 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KAT6A | NM_001305878.1 | NP_001292807.1 | ||
| NM_006766.4 | NP_006757.2 | |||
| XM_011544656.2 | XP_011542958.1 | |||
| XM_011544657.2 | XP_011542959.1 | |||
| XM_011544658.2 | XP_011542960.1 | |||
| XM_011544659.2 | XP_011542961.1 | |||
| XM_017013863.1 | XP_016869352.1 | |||
| XM_017013864.1 | XP_016869353.1 | |||
| BC142659.1 | ||||
| BC142959.1 | ||||
| BP248857.1 | ||||
| BP271264.1 | ||||
| BP348860.1 | ||||
| DA257528.1 | ||||
| DA442995.1 | ||||
| DA758316.1 | ||||
| U47742.1 | AAC50662.1 |
Target Gene Details
Entrez Gene ID: | 105379393 |
Gene Name: | uncharacterized LOC105379393 |
Gene Aliases: |
- |
Location: |
Chr.8:42051750-42104987 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105379393 | NR_136589.1 | |||
| HY030269.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521189 | Chr.8:42050630 - 42056486 on Build GRCh38 | Loss |
|
| nsv824615 | Chr.8:42049114 - 42054345 on Build GRCh38 | Gain |
|
| nsv1112962 | Chr.8:42051282 - 42051982 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map