Assay Details
Target Gene Details
Entrez Gene ID: | 101929402 |
Gene Name: | EXTL3 antisense RNA 1 |
Gene Aliases: |
C8orf50 |
Location: |
Chr.8:28698403-28701464 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EXTL3-AS1 | NR_126027.1 |

Genomic Map