Assay Details
Target Gene Details
Entrez Gene ID: | 105375938 |
Gene Name: | translation initiation factor IF-2-like |
Gene Aliases: |
- |
Location: |
Chr.8:60909471-60910926 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105375938 | XM_011517657.2 | 2 | 872 | XP_011515959.1 |
| XM_017014120.1 | 2 | 875 | XP_016869609.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526196 | Chr.8:60681010 - 60965064 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs112719526] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map