Assay Details
Target Gene Details
Entrez Gene ID: | 7013 |
Gene Name: | telomeric repeat binding factor 1 |
Gene Aliases: |
PIN2, TRBF1, TRF, TRF1, hTRF1-AS, t-TRF1 |
Location: |
Chr.8:73008862-73047752 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TERF1 | NM_003218.3 | NP_003209.2 | ||
| NM_017489.2 | NP_059523.2 | |||
| XM_005251291.3 | XP_005251348.1 | |||
| XM_005251292.3 | XP_005251349.1 | |||
| XM_011517582.2 | XP_011515884.1 | |||
| XM_017013792.1 | XP_016869281.1 | |||
| AF003001.1 | AAB81137.1 | |||
| AK292529.1 | ||||
| BC029378.1 | AAH29378.1 | |||
| U40705.1 | AAB54036.1 | |||
| U74382.1 | AAB53363.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831354 | Chr.8:72906156 - 73065096 on Build GRCh38 | Gain |
|
| esv2737154 | Chr.8:71825978 - 73469084 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79263711] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map