Assay Details
Target Gene Details
Entrez Gene ID: | 5440 |
Gene Name: | RNA polymerase II subunit K |
Gene Aliases: |
ABC10-alpha, RPABC4, RPB10alpha, RPB12, RPB7.0, hRPB7.0, hsRPB10a |
Location: |
Chr.8:100150611-100154002 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLR2K | NM_005034.3 | NP_005025.1 | ||
| BC000806.1 | AAH00806.1 | |||
| BC018157.1 | AAH18157.1 | |||
| CR456870.1 | CAG33151.1 | |||
| Z47727.1 | CAA87656.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv611814 | Chr.8:100097796 - 100177024 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map