Assay Details
Target Gene Details
Entrez Gene ID: | 8756 |
Gene Name: | ADAM metallopeptidase domain 7 |
Gene Aliases: |
ADAM 7, ADAM-7, EAPI, GP-83, GP83 |
Location: |
Chr.8:24440996-24509565 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAM7 | NM_003817.3 | NP_003808.2 | ||
| XM_017013943.1 | XP_016869432.1 | |||
| AF090327.1 | AAC36742.1 | |||
| AF215824.1 | AAG43987.1 | |||
| AK292492.1 | ||||
| BC043207.2 | AAH43207.2 |
Target Gene Details
Entrez Gene ID: | 101929294 |
Gene Name: | uncharacterized LOC101929294 |
Gene Aliases: |
- |
Location: |
Chr.8:24295814-24548618 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929294 | NR_125808.1 | |||
| DA221074.1 |
Target Gene Details
Entrez Gene ID: | 101929315 |
Gene Name: | uncharacterized LOC101929315 |
Gene Aliases: |
- |
Location: |
Chr.8:24490312-24514868 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929315 | NR_125809.1 | |||
| BX096372.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891377 | Chr.8:23722497 - 25043041 on Build GRCh38 | Loss |
|
| dgv452n21 | Chr.8:24421431 - 24505297 on Build GRCh38 | Loss |
|
| dgv7141n100 | Chr.8:24436486 - 24511427 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map