Assay Details
Target Gene Details
Entrez Gene ID: | 105375650 |
Gene Name: | uncharacterized LOC105375650 |
Gene Aliases: |
- |
Location: |
Chr.8:95066808-95073182 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105375650 | NR_132986.1 | 2 | 603 | |
| NR_132987.1 | 2 | 553 |
Target Gene Details
Entrez Gene ID: | 137682 |
Gene Name: | NADH:ubiquinone oxidoreductase complex assembly factor 6 |
Gene Aliases: |
C8orf38 |
Location: |
Chr.8:94895271-95118496 on Build GRCh38 |
Assay Gene Location: | Within Intron 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFAF6 | XM_017013039.1 | XP_016868528.1 | ||
| AK301414.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508520 | Chr.8:94927145 - 95127840 on Build GRCh38 | Deletion |
|
| esv2667210 | Chr.8:95067174 - 95069951 on Build GRCh38 | Deletion |
|
| esv24383 | Chr.8:95067076 - 95069463 on Build GRCh38 | Loss |
|
| esv3618097 | Chr.8:95067153 - 95070097 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map