Assay Details
Target Gene Details
Entrez Gene ID: | 79648 |
Gene Name: | microcephalin 1 |
Gene Aliases: |
BRIT1, MCT |
Location: |
Chr.8:6406592-6648505 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv527005 | Chr.8:6271530 - 6429649 on Build GRCh38 | Gain |
|
| dgv6868n100 | Chr.8:6266604 - 6447293 on Build GRCh38 | Gain |
|
| nsv524404 | Chr.8:6405562 - 6433964 on Build GRCh38 | Loss |
|
| nsv1018782 | Chr.8:6126510 - 6426852 on Build GRCh38 | Loss |
|
| nsv1030323 | Chr.8:6410707 - 6421990 on Build GRCh38 | Loss |
|
| esv3576515 | Chr.8:6377962 - 6478027 on Build GRCh38 | Gain |
|
| nsv1018882 | Chr.8:6347336 - 6432416 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map