Assay Details
Target Gene Details
Entrez Gene ID: | 105379393 |
Gene Name: | uncharacterized LOC105379393 |
Gene Aliases: |
- |
Location: |
Chr.8:42051750-42104987 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105379393 | NR_136589.1 | |||
| HY030269.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6168 | Chr.8:42092972 - 42137630 on Build GRCh38 | Deletion |
|
| nsv1022636 | Chr.8:42093555 - 42134046 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs150947192] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map