Assay Details
Target Gene Details
Entrez Gene ID: | 28998 |
Gene Name: | mitochondrial ribosomal protein L13 |
Gene Aliases: |
L13, L13A, L13mt, RPL13, RPML13 |
Location: |
Chr.8:120395844-120445407 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL13 | NM_014078.5 | NP_054797.2 | ||
| AB049640.1 | BAB40845.1 | |||
| AF112214.1 | AAF17202.1 | |||
| AK307322.1 | ||||
| AK311924.1 | ||||
| BC009190.2 | AAH09190.1 | |||
| BC021744.2 | AAH21744.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470235 | Chr.8:120424982 - 120514507 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map