Assay Details
Target Gene Details
Entrez Gene ID: | 6674 |
Gene Name: | sperm associated antigen 1 |
Gene Aliases: |
CILD28, CT140, HEL-S-268, HSD-3.8, SP75, TPIS |
Location: |
Chr.8:100157869-100241904 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPAG1 | XM_017013754.1 | XP_016869243.1 | ||
| XM_017013755.1 | XP_016869244.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv611814 | Chr.8:100097796 - 100177024 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map