Assay Details
Target Gene Details
Entrez Gene ID: | 10499 |
Gene Name: | nuclear receptor coactivator 2 |
Gene Aliases: |
GRIP1, KAT13C, NCoA-2, SRC2, TIF2, bHLHe75 |
Location: |
Chr.8:70109762-70405390 on Build GRCh38 |
Assay Gene Location: | Within Intron 27 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831352 | Chr.8:69999548 - 70182606 on Build GRCh38 | Gain |
|
| nsv6246 | Chr.8:70106516 - 70137048 on Build GRCh38 | Insertion |
|
| nsv6247 | Chr.8:70118114 - 70163017 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74982167] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map