Assay Details
Target Gene Details
Entrez Gene ID: | 157869 |
Gene Name: | somatomedin B and thrombospondin type 1 domain containing |
Gene Aliases: |
C8orf84, RPESP |
Location: |
Chr.8:73063077-73093272 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SBSPON | NM_153225.3 | 5 | 2018 | NP_694957.3 |
| XM_017013145.1 | 5 | 2263 | XP_016868634.1 | |
| AF087977.1 | 1 | 68 | ||
| AL080094.1 | 1 | 630 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2737154 | Chr.8:71825978 - 73469084 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map